Assay Details
| Gene Symbol: Prss33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864665.3 | XP_004864722.2 | 3 - 4 | 206 | 63 | |
| XM_004864666.3 | XP_004864723.2 | 3 - 4 | 206 | 63 | ||
| XM_004864667.3 | XP_004864724.1 | 3 - 4 | 206 | 63 | ||
| XM_004864668.2 | XP_004864725.1 | 2 - 3 | 136 | 63 | ||
| XM_004864670.3 | XP_004864727.2 | 3 - 4 | 206 | 63 | ||
| XM_013065642.2 | XP_012921096.1 | 3 - 4 | 124 | 63 | ||
| XM_021244389.1 | XP_021100048.1 | 2 - 3 | 92 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021244389.1 transcripts.
Set Membership: |
|

Prss33
Genomic Map