Assay Details
| Gene Symbol: Mypop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867124.2 | XP_004867181.1 | 2 - 3 | 1930 | 78 | |
| XM_021245872.1 | XP_021101531.1 | 3 - 4 | 1256 | 78 | ||
| XM_021245873.1 | XP_021101532.1 | 3 - 4 | 1467 | 78 | ||
More Information
Set Membership: |
|

Mypop
Genomic Map