Assay Details
Gene Symbol: Cnih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867458.2 | XP_004867515.1 | 5 - 6 | 536 | 72 | |
XM_004867460.2 | XP_004867517.1 | 4 - 5 | 488 | 72 | ||
XM_013066543.2 | XP_012921997.1 | 7 - 8 | 993 | 72 | ||
XM_021246455.1 | XP_021102114.1 | 7 - 8 | 1933 | 72 | ||
XM_021246456.1 | XP_021102115.1 | 7 - 8 | 1226 | 72 | ||
XM_021246457.1 | XP_021102116.1 | 6 - 7 | 818 | 72 | ||
XM_021246458.1 | XP_021102117.1 | 6 - 7 | 1682 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
