Assay Details
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867675.3 | XP_004867732.1 | 4 - 5 | 681 | 84 | |
XM_021246677.1 | XP_021102336.1 | 6 - 7 | 827 | 84 | ||
XM_021246678.1 | XP_021102337.1 | 5 - 6 | 544 | 84 | ||
XM_021246679.1 | XP_021102338.1 | 4 - 5 | 624 | 84 | ||
XM_021246680.1 | XP_021102339.1 | 2 - 3 | 1201 | 84 | ||
XM_021246683.1 | XP_021102342.1 | 2 - 3 | 311 | 84 |
