Assay Details
| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870759.2 | XP_004870816.1 | 5 - 6 | 1308 | 68 | |
| XM_004870760.2 | XP_004870817.1 | 4 - 5 | 1217 | 68 | ||
More Information
Set Membership: |
|

Trim16
Genomic Map