Assay Details
Gene Symbol: LOC101710340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068106.2 | XP_012923560.1 | 18 - 19 | 2143 | 93 | |
XM_021249801.1 | XP_021105460.1 | 18 - 19 | 2143 | 93 | ||
XM_021249802.1 | XP_021105461.1 | 18 - 19 | 2144 | 93 |
