Assay Details
| Gene Symbol: LOC101711022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245134.1 | XP_021100793.1 | 4 - 5 | 1699 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101711022| Gene Symbol: LOC101711022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245134.1 | XP_021100793.1 | 4 - 5 | 1699 | 88 | |
Set Membership: |
|