Assay Details
| Gene Symbol: Mageb17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245360.1 | XP_021101019.1 | 1 - 2 | 47 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021245360.1 transcripts.
Set Membership: |
|

Mageb17
Genomic Map