Assay Details
| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255249.1 | XP_021110908.1 | 3 - 4 | 3966 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srms| Gene Symbol: Srms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255249.1 | XP_021110908.1 | 3 - 4 | 3966 | 55 | |
Set Membership: |
|