Assay Details
| Gene Symbol: LOC101699075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256826.1 | XP_021112485.1 | 4 - 5 | 486 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101699075| Gene Symbol: LOC101699075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256826.1 | XP_021112485.1 | 4 - 5 | 486 | 70 | |
Set Membership: |
|