Assay Details
| Gene Symbol: Rfx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835343.2 | XP_004835400.1 | 6 - 7 | 522 | 65 | |
| XM_021260701.1 | XP_021116360.1 | 6 - 7 | 910 | 65 | ||
More Information
Set Membership: |
|

Rfx7
Genomic Map