Assay Details
| Gene Symbol: Ccng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836427.2 | XP_004836484.1 | 5 - 6 | 824 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccng1| Gene Symbol: Ccng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836427.2 | XP_004836484.1 | 5 - 6 | 824 | 75 | |
Set Membership: |
|