Assay Details
| Gene Symbol: Ppp1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842093.2 | XP_004842150.2 | 5 - 6 | 775 | 92 | |
| XM_021256059.1 | XP_021111718.1 | 5 - 6 | 721 | 92 | ||
More Information
Set Membership: |
|

Ppp1r42
Genomic Map