Assay Details
| Gene Symbol: LOC101712048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851520.3 | XP_004851577.1 | 1 - 2 | 206 | 91 | |
| XM_004851523.3 | XP_004851580.1 | 1 - 2 | 206 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004851520.3, XM_004851523.3 transcripts.
Set Membership: |
|

LOC101712048
Genomic Map