Assay Details
| Gene Symbol: LOC101712048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851520.3 | XP_004851577.1 | 2 - 3 | 304 | 66 | |
| XM_004851521.3 | XP_004851578.1 | 2 - 3 | 253 | 66 | ||
| XM_004851522.3 | XP_004851579.1 | 2 - 3 | 301 | 66 | ||
| XM_004851523.3 | XP_004851580.1 | 2 - 3 | 304 | 66 | ||
More Information
Set Membership: |
|

LOC101712048
Genomic Map