Assay Details
| Gene Symbol: LOC101712048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851520.3 | XP_004851577.1 | 3 - 4 | 441 | 91 | |
| XM_004851521.3 | XP_004851578.1 | 3 - 4 | 390 | 91 | ||
| XM_004851522.3 | XP_004851579.1 | 3 - 4 | 438 | 91 | ||
More Information
Set Membership: |
|

LOC101712048
Genomic Map