Assay Details
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852210.3 | XP_004852267.1 | 7 - 8 | 1021 | 51 | |
XM_004852211.3 | XP_004852268.1 | 7 - 8 | 908 | 51 | ||
XM_013075029.2 | XP_012930483.1 | 7 - 8 | 1006 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
