Assay Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857627.2 | XP_004857684.1 | 5 - 6 | 1799 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl41| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857627.2 | XP_004857684.1 | 5 - 6 | 1799 | 68 | |
Set Membership: |
|