Assay Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858014.2 | XP_004858071.1 | 1 - 2 | 187 | 87 | |
| XM_021238929.1 | XP_021094588.1 | 1 - 2 | 187 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004858014.2, XM_021238929.1 transcripts.
Set Membership: |
|

Cass4
Genomic Map