Assay Details
| Gene Symbol: Kcng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858045.3 | XP_004858102.1 | 2 - 3 | 1019 | 80 | |
| XM_021239047.1 | XP_021094706.1 | 2 - 3 | 1775 | 80 | ||
| XM_021239048.1 | XP_021094707.1 | 1 - 2 | 782 | 80 | ||
More Information
Set Membership: |
|

Kcng1
Genomic Map