Assay Details
| Gene Symbol: Coasy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859326.3 | XP_004859383.1 | 7 - 8 | 1707 | 64 | |
| XM_013077728.2 | XP_012933182.1 | 7 - 8 | 1717 | 64 | ||
More Information
Set Membership: |
|

Coasy
Genomic Map