Assay Details
| Gene Symbol: Llgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860683.3 | XP_004860740.1 | 2 - 3 | 301 | 57 | |
| XM_013078113.2 | XP_012933567.1 | 2 - 3 | 294 | 57 | ||
| XM_013078114.2 | XP_012933568.1 | 2 - 3 | 295 | 57 | ||
| XM_021241090.1 | XP_021096749.1 | 2 - 3 | 305 | 57 | ||
| XM_021241091.1 | XP_021096750.1 | 2 - 3 | 299 | 57 | ||
| XM_021241092.1 | XP_021096751.1 | 2 - 3 | 295 | 57 | ||
| XM_021241094.1 | XP_021096753.1 | 2 - 3 | 244 | 57 | ||
| XM_021241095.1 | XP_021096754.1 | 3 - 4 | 442 | 57 | ||
| XM_021241096.1 | XP_021096755.1 | 2 - 3 | 122 | 57 | ||
More Information
Set Membership: |
|

Llgl2
Genomic Map