Assay Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861050.3 | XP_004861107.1 | 3 - 4 | 1034 | 65 | |
| XM_013078116.2 | XP_012933570.1 | 3 - 4 | 1034 | 65 | ||
| XM_013078117.2 | XP_012933571.1 | 4 - 5 | 1163 | 65 | ||
| XM_013078119.2 | XP_012933573.1 | 3 - 4 | 1034 | 65 | ||
| XM_013078120.2 | XP_012933574.1 | 3 - 4 | 1034 | 65 | ||
| XM_021241303.1 | XP_021096962.1 | 3 - 4 | 1034 | 65 | ||
| XM_021241304.1 | XP_021096963.1 | 3 - 4 | 1034 | 65 | ||
| XM_021241305.1 | XP_021096964.1 | 3 - 4 | 872 | 65 | ||
| XM_021241306.1 | XP_021096965.1 | 3 - 4 | 965 | 65 | ||
| XM_021241307.1 | XP_021096966.1 | 3 - 4 | 1034 | 65 | ||
| XM_021241309.1 | XP_021096968.1 | 3 - 4 | 1034 | 65 | ||
| XM_021241310.1 | XP_021096969.1 | 3 - 4 | 1034 | 65 | ||
More Information
Set Membership: |
|

B3gntl1
Genomic Map