Assay Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861057.2 | XP_004861114.1 | 5 - 6 | 887 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fn3k| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861057.2 | XP_004861114.1 | 5 - 6 | 887 | 62 | |
Set Membership: |
|