Assay Details
| Gene Symbol: Crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865621.3 | XP_004865678.2 | 7 - 8 | 860 | 72 | |
| XM_004865622.3 | XP_004865679.2 | 7 - 8 | 851 | 72 | ||
| XM_004865623.3 | XP_004865680.2 | 6 - 7 | 803 | 72 | ||
| XM_004865624.3 | XP_004865681.1 | 5 - 6 | 476 | 72 | ||
| XM_021245135.1 | XP_021100794.1 | 3 - 4 | 344 | 72 | ||
| XM_021245136.1 | XP_021100795.1 | 6 - 7 | 622 | 72 | ||
More Information
Set Membership: |
|

Crb3
Genomic Map