Assay Details
| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870690.3 | XP_004870747.1 | 3 - 4 | 591 | 55 | |
| XM_021248881.1 | XP_021104540.1 | 3 - 4 | 780 | 55 | ||
More Information
Set Membership: |
|

Rnf135
Genomic Map