Assay Details
| Gene Symbol: Armcx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872565.3 | XP_004872622.1 | 4 - 5 | 443 | 124 | |
| XM_004872566.3 | XP_004872623.1 | 3 - 4 | 354 | 124 | ||
| XM_021250455.1 | XP_021106114.1 | 4 - 5 | 407 | 124 | ||
| XM_021250457.1 | XP_021106116.1 | 3 - 4 | 318 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004872565.3, XM_004872566.3, XM_021250455.1, XM_021250457.1 transcripts.
Set Membership: |
|

Armcx4
Genomic Map