Assay Details
| Gene Symbol: Znf567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077418.2 | XP_012932872.1 | 5 - 6 | 1544 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf567| Gene Symbol: Znf567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077418.2 | XP_012932872.1 | 5 - 6 | 1544 | 108 | |
Set Membership: |
|