Assay Details
| Gene Symbol: Cd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251863.1 | XP_021107522.1 | 8 - 9 | 1715 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd5| Gene Symbol: Cd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251863.1 | XP_021107522.1 | 8 - 9 | 1715 | 56 | |
Set Membership: |
|