Assay Details
| Gene Symbol: LOC101717885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021264527.1 | XP_021120186.1 | 3 - 4 | 1118 | 70 | |
| XM_021264530.1 | XP_021120189.1 | 3 - 4 | 1243 | 70 | ||
| XM_021264531.1 | XP_021120190.1 | 3 - 4 | 1140 | 70 | ||
| XM_021264532.1 | XP_021120191.1 | 3 - 4 | 1397 | 70 | ||
More Information
Set Membership: |
|

LOC101717885
Genomic Map