Assay Details
| Gene Symbol: Faxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836510.3 | XP_004836567.1 | 3 - 4 | 722 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Faxdc2| Gene Symbol: Faxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836510.3 | XP_004836567.1 | 3 - 4 | 722 | 58 | |
Set Membership: |
|