Assay Details
| Gene Symbol: Nfe2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839531.3 | XP_004839588.1 | 1 - 2 | 638 | 114 | |
| XM_021254103.1 | XP_021109762.1 | 1 - 2 | 638 | 114 | ||
More Information
Set Membership: |
|

Nfe2l3
Genomic Map