Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 11 - 12 | 2277 | 99 | |
| XM_021254747.1 | XP_021110406.1 | 11 - 12 | 2280 | 99 | ||
| XM_021254748.1 | XP_021110407.1 | 11 - 12 | 2244 | 99 | ||
| XM_021254750.1 | XP_021110409.1 | 11 - 12 | 2251 | 99 | ||
| XM_021254751.1 | XP_021110410.1 | 11 - 12 | 2226 | 99 | ||
| XM_021254752.1 | XP_021110411.1 | 11 - 12 | 2283 | 99 | ||
| XM_021254753.1 | XP_021110412.1 | 11 - 12 | 2258 | 99 | ||
| XM_021254754.1 | XP_021110413.1 | 10 - 11 | 2049 | 99 | ||
| XM_021254755.1 | XP_021110414.1 | 11 - 12 | 2279 | 99 | ||
| XM_021254756.1 | XP_021110415.1 | 8 - 9 | 2612 | 99 | ||
| XM_021254757.1 | XP_021110416.1 | 7 - 8 | 1305 | 99 | ||
| XM_021254758.1 | XP_021110417.1 | 11 - 12 | 2225 | 99 | ||

LOC101713044
Genomic Map