Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 14 - 15 | 2614 | 100 | |
| XM_021254747.1 | XP_021110406.1 | 14 - 15 | 2617 | 100 | ||
| XM_021254748.1 | XP_021110407.1 | 14 - 15 | 2581 | 100 | ||
| XM_021254750.1 | XP_021110409.1 | 14 - 15 | 2588 | 100 | ||
| XM_021254751.1 | XP_021110410.1 | 14 - 15 | 2563 | 100 | ||
| XM_021254752.1 | XP_021110411.1 | 14 - 15 | 2620 | 100 | ||
| XM_021254753.1 | XP_021110412.1 | 14 - 15 | 2595 | 100 | ||
| XM_021254754.1 | XP_021110413.1 | 13 - 14 | 2386 | 100 | ||
| XM_021254755.1 | XP_021110414.1 | 14 - 15 | 2616 | 100 | ||
| XM_021254756.1 | XP_021110415.1 | 11 - 12 | 2949 | 100 | ||
| XM_021254757.1 | XP_021110416.1 | 10 - 11 | 1642 | 100 | ||
| XM_021254758.1 | XP_021110417.1 | 14 - 15 | 2562 | 100 | ||

LOC101713044
Genomic Map