Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 18 - 19 | 3080 | 75 | |
| XM_021254747.1 | XP_021110406.1 | 18 - 19 | 3083 | 75 | ||
| XM_021254748.1 | XP_021110407.1 | 18 - 19 | 3047 | 75 | ||
| XM_021254750.1 | XP_021110409.1 | 18 - 19 | 3054 | 75 | ||
| XM_021254751.1 | XP_021110410.1 | 18 - 19 | 3029 | 75 | ||
| XM_021254752.1 | XP_021110411.1 | 17 - 18 | 2960 | 75 | ||
| XM_021254753.1 | XP_021110412.1 | 18 - 19 | 3061 | 75 | ||
| XM_021254754.1 | XP_021110413.1 | 17 - 18 | 2852 | 75 | ||
| XM_021254756.1 | XP_021110415.1 | 15 - 16 | 3415 | 75 | ||
| XM_021254757.1 | XP_021110416.1 | 14 - 15 | 2108 | 75 | ||
| XM_021254758.1 | XP_021110417.1 | 18 - 19 | 3028 | 75 | ||
More Information
Set Membership: |
|

LOC101713044
Genomic Map