Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 4 - 5 | 1194 | 77 | |
| XM_021254747.1 | XP_021110406.1 | 4 - 5 | 1197 | 77 | ||
| XM_021254748.1 | XP_021110407.1 | 4 - 5 | 1161 | 77 | ||
| XM_021254750.1 | XP_021110409.1 | 4 - 5 | 1168 | 77 | ||
| XM_021254751.1 | XP_021110410.1 | 4 - 5 | 1197 | 77 | ||
| XM_021254752.1 | XP_021110411.1 | 4 - 5 | 1200 | 77 | ||
| XM_021254753.1 | XP_021110412.1 | 4 - 5 | 1175 | 77 | ||
| XM_021254754.1 | XP_021110413.1 | 4 - 5 | 1197 | 77 | ||
| XM_021254755.1 | XP_021110414.1 | 4 - 5 | 1196 | 77 | ||
| XM_021254758.1 | XP_021110417.1 | 4 - 5 | 1196 | 77 | ||

LOC101713044
Genomic Map