Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 8 - 9 | 1805 | 76 | |
| XM_021254747.1 | XP_021110406.1 | 8 - 9 | 1808 | 76 | ||
| XM_021254748.1 | XP_021110407.1 | 8 - 9 | 1772 | 76 | ||
| XM_021254750.1 | XP_021110409.1 | 8 - 9 | 1779 | 76 | ||
| XM_021254751.1 | XP_021110410.1 | 8 - 9 | 1754 | 76 | ||
| XM_021254752.1 | XP_021110411.1 | 8 - 9 | 1811 | 76 | ||
| XM_021254753.1 | XP_021110412.1 | 8 - 9 | 1786 | 76 | ||
| XM_021254755.1 | XP_021110414.1 | 8 - 9 | 1807 | 76 | ||
| XM_021254756.1 | XP_021110415.1 | 5 - 6 | 2140 | 76 | ||
| XM_021254757.1 | XP_021110416.1 | 4 - 5 | 833 | 76 | ||
| XM_021254758.1 | XP_021110417.1 | 8 - 9 | 1753 | 76 | ||

LOC101713044
Genomic Map