Assay Details
| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849182.2 | XP_004849239.1 | 2 - 3 | 773 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Endog| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849182.2 | XP_004849239.1 | 2 - 3 | 773 | 66 | |
Set Membership: |
|