Assay Details
| Gene Symbol: Mocos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855231.2 | XP_004855288.1 | 13 - 14 | 2481 | 70 | |
| XM_021236686.1 | XP_021092345.1 | 13 - 14 | 3038 | 70 | ||
| XM_021236687.1 | XP_021092346.1 | 12 - 13 | 2971 | 70 | ||
More Information
Set Membership: |
|

Mocos
Genomic Map