Assay Details
| Gene Symbol: Syt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866677.3 | XP_004866734.1 | 3 - 4 | 345 | 57 | |
| XM_004866678.3 | XP_004866735.1 | 3 - 4 | 345 | 57 | ||
More Information
Set Membership: |
|

Syt5
Genomic Map