Assay Details
Gene Symbol: Bbc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867068.2 | XP_004867125.1 | 2 - 3 | 370 | 115 | |
XM_004867069.3 | XP_004867126.1 | 2 - 3 | 366 | 115 | ||
XM_004867070.3 | XP_004867127.1 | 2 - 3 | 365 | 115 | ||
XM_004867071.3 | XP_004867128.1 | 2 - 3 | 357 | 115 | ||
XM_004867072.3 | XP_004867129.1 | 2 - 3 | 318 | 115 | ||
XM_021245699.1 | XP_021101358.1 | 2 - 3 | 406 | 115 | ||
XM_021245700.1 | XP_021101359.1 | 2 - 3 | 335 | 115 |

More Information
Set Membership: |
![]() ![]() |
