Assay Details
| Gene Symbol: LOC101720177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867430.3 | XP_004867487.1 | 17 - 18 | 1952 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101720177| Gene Symbol: LOC101720177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867430.3 | XP_004867487.1 | 17 - 18 | 1952 | 67 | |
Set Membership: |
|