Assay Details
| Gene Symbol: Pld6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868737.2 | XP_004868794.1 | 1 - 2 | 458 | 91 | |
| XM_013067041.2 | XP_012922495.1 | 3 - 4 | 681 | 91 | ||
More Information
Set Membership: |
|

Pld6
Genomic Map