Assay Details
| Gene Symbol: Zfp91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869742.3 | XP_004869799.1 | 7 - 8 | 1024 | 72 | |
| XM_021248343.1 | XP_021104002.1 | 7 - 8 | 1039 | 72 | ||
| XM_021248344.1 | XP_021104003.1 | 7 - 8 | 1036 | 72 | ||
| XM_021248345.1 | XP_021104004.1 | 7 - 8 | 1022 | 72 | ||
| XM_021248346.1 | XP_021104005.1 | 7 - 8 | 771 | 72 | ||
More Information
Set Membership: |
|

Zfp91
Genomic Map