Assay Details
| Gene Symbol: Nt5c3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872195.2 | XP_004872252.1 | 7 - 8 | 742 | 91 | |
| XM_004872196.3 | XP_004872253.1 | 7 - 8 | 2487 | 91 | ||
| XM_004872197.3 | XP_004872254.1 | 8 - 9 | 1731 | 91 | ||
| XM_004872198.3 | XP_004872255.1 | 8 - 9 | 718 | 91 | ||
| XM_004872199.3 | XP_004872256.1 | 8 - 9 | 716 | 91 | ||
| XM_013068302.2 | XP_012923756.1 | 9 - 10 | 856 | 91 | ||
| XM_021250156.1 | XP_021105815.1 | 9 - 10 | 1898 | 91 | ||
More Information
Set Membership: |
|

Nt5c3a
Genomic Map