Assay Details
| Gene Symbol: Csgalnact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874097.3 | XP_004874154.1 | 4 - 5 | 1746 | 70 | |
| XM_004874098.3 | XP_004874155.1 | 3 - 4 | 1657 | 70 | ||
| XM_021251615.1 | XP_021107274.1 | 4 - 5 | 1746 | 70 | ||
| XM_021251616.1 | XP_021107275.1 | 4 - 5 | 1746 | 70 | ||
More Information
Set Membership: |
|

Csgalnact2
Genomic Map