Assay Details
| Gene Symbol: Bsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066208.2 | XP_012921662.2 | 4 - 5 | 222 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bsph1| Gene Symbol: Bsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066208.2 | XP_012921662.2 | 4 - 5 | 222 | 81 | |