Assay Details
Gene Symbol: LOC101714324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068852.2 | XP_012924306.1 | 18 - 19 | 4452 | 105 | |
XM_021249578.1 | XP_021105237.1 | 18 - 19 | 4449 | 105 | ||
XM_021249580.1 | XP_021105239.1 | 18 - 19 | 4450 | 105 | ||
XM_021249582.1 | XP_021105241.1 | 17 - 18 | 4367 | 105 | ||
XM_021249588.1 | XP_021105247.1 | 18 - 19 | 4286 | 105 | ||
XM_021249597.1 | XP_021105256.1 | 18 - 19 | 4283 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
