Assay Details
| Gene Symbol: CUNH21orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071395.2 | XP_012926849.1 | 1 - 2 | 1926 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH21orf58| Gene Symbol: CUNH21orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071395.2 | XP_012926849.1 | 1 - 2 | 1926 | 64 | |
Set Membership: |
|