Assay Details
| Gene Symbol: Malt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239239.1 | XP_021094898.1 | 12 - 13 | 1668 | 74 | |
| XM_021239240.1 | XP_021094899.1 | 11 - 12 | 1635 | 74 | ||
More Information
Set Membership: |
|

Malt1
Genomic Map